Assay Details
Gene Symbol: lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201306.1 | NP_958463.1 | 2 - 3 | 408 | 69 | |
GenBank mRNA | BC049517.1 | - | 2 - 3 | 408 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
