Assay Details
| Gene Symbol: fn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013261.1 | NP_001013279.1 | 10 - 11 | 1648 | 69 | |
| XM_005160284.3 | XP_005160341.1 | 10 - 11 | 1683 | 69 | ||
| XM_009305554.3 | XP_009303829.1 | 10 - 11 | 1684 | 69 | ||
| GenBank mRNA | AY538257.1 | - | 10 - 11 | 1648 | 69 | |
| AY725818.1 | - | 10 - 11 | 1663 | 69 | ||
More Information
Set Membership: |
|

fn1b
Genomic Map