Assay Details
| Gene Symbol: rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024819.1 | NP_001019990.1 | 4 - 5 | 692 | 58 | |
| GenBank mRNA | AY613958.1 | - | 4 - 5 | 692 | 58 | |
More Information
Set Membership: |
|

rhd
Genomic Map