Assay Details
| Gene Symbol: wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030261.2 | NP_001025432.2 | 3 - 4 | 629 | 56 | |
| XM_021468895.1 | XP_021324570.1 | 2 - 3 | 534 | 56 | ||
| GenBank mRNA | BC098539.1 | - | 3 - 4 | 629 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021468895.1 transcripts.
Set Membership: |
|

wdr41
Genomic Map