Assay Details
| Gene Symbol: cluha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077560.1 | NP_001071028.1 | 10 - 11 | 2149 | 68 | |
| XM_005157586.4 | XP_005157643.1 | 10 - 11 | 2196 | 68 | ||
| XM_005157587.4 | XP_005157644.1 | 10 - 11 | 1952 | 68 | ||
| XM_021481439.1 | XP_021337114.1 | 10 - 11 | 2196 | 68 | ||
| GenBank mRNA | BC125829.1 | - | 10 - 11 | 2149 | 68 | |
More Information
Set Membership: |
|

cluha
Genomic Map