Assay Details
| Gene Symbol: st14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077737.1 | NP_001071205.1 | 6 - 7 | 921 | 65 | |
| XM_005161204.4 | XP_005161261.1 | 7 - 8 | 877 | 65 | ||
| XM_009295332.3 | XP_009293607.1 | 7 - 8 | 797 | 65 | ||
| GenBank mRNA | BC125837.1 | - | 6 - 7 | 921 | 65 | |
| BC165661.1 | - | 6 - 7 | 708 | 65 | ||
More Information
Set Membership: |
|

st14b
Genomic Map