Assay Details
| Gene Symbol: tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079987.2 | NP_001073456.1 | 1 - 2 | 244 | 86 | |
| GenBank mRNA | BC128830.1 | - | 1 - 2 | 245 | 86 | |
More Information
Set Membership: |
|

tmem41b
Genomic Map