Assay Details
| Gene Symbol: gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199655.1 | NP_955949.1 | 10 - 11 | 1279 | 67 | |
| GenBank mRNA | AY391428.1 | - | 10 - 11 | 1203 | 67 | |
| BC045493.1 | - | 10 - 11 | 1279 | 67 | ||
More Information
Set Membership: |
|

gdi2
Genomic Map