Assay Details
| Gene Symbol: scp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200865.2 | NP_957159.1 | 4 - 5 | 407 | 58 | |
| GenBank mRNA | BC063230.1 | - | 4 - 5 | 341 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scp2a| Gene Symbol: scp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200865.2 | NP_957159.1 | 4 - 5 | 407 | 58 | |
| GenBank mRNA | BC063230.1 | - | 4 - 5 | 341 | 58 | |
Set Membership: |
|