Assay Details
| Gene Symbol: scp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200865.2 | NP_957159.1 | 5 - 6 | 470 | 100 | |
| GenBank mRNA | BC063230.1 | - | 5 - 6 | 404 | 100 | |
More Information
Set Membership: |
|

scp2a
Genomic Map