Assay Details
| Gene Symbol: h2ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201073.1 | NP_957367.1 | 1 | 542 | 86 | |
| GenBank mRNA | BC046078.1 | - | 1 | 542 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

h2ax| Gene Symbol: h2ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201073.1 | NP_957367.1 | 1 | 542 | 86 | |
| GenBank mRNA | BC046078.1 | - | 1 | 542 | 86 | |
Set Membership: |
|