Assay Details
| Gene Symbol: sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001816.4 | NP_001001816.2 | 6 - 7 | 868 | 56 | |
| XM_009295594.3 | XP_009293869.1 | 6 - 7 | 1027 | 56 | ||
| GenBank mRNA | AY398418.1 | - | 5 - 6 | 578 | 56 | |
| BC095349.1 | - | 6 - 7 | 868 | 56 | ||
| BC164654.1 | - | 5 - 6 | 606 | 56 | ||
More Information
Set Membership: |
|

sgcd
Genomic Map