Assay Details
| Gene Symbol: mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076565.1 | NP_001070033.1 | 3 - 4 | 533 | 73 | |
| GenBank mRNA | BC124239.1 | - | 3 - 4 | 533 | 73 | |
More Information
Set Membership: |
|

mpnd
Genomic Map