Assay Details
| Gene Symbol: bag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098736.2 | NP_001092206.1 | 5 - 6 | 545 | 75 | |
| GenBank mRNA | BC142844.1 | - | 5 - 6 | 487 | 75 | |
| EH437526.1 | - | 5 - 6 | 557 | 75 | ||
More Information
Set Membership: |
|

bag1
Genomic Map