Assay Details
| Gene Symbol: paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199537.2 | NP_955831.1 | 5 - 6 | 851 | 62 | |
| GenBank mRNA | BC048051.1 | - | 5 - 6 | 814 | 62 | |
| BC071527.1 | - | 5 - 6 | 851 | 62 | ||
More Information
Set Membership: |
|

paics
Genomic Map