Assay Details
| Gene Symbol: dcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200152.1 | NP_956446.1 | 2 - 3 | 405 | 63 | |
| GenBank mRNA | BC044547.1 | - | 2 - 3 | 405 | 63 | |
| BC066577.1 | - | 2 - 3 | 436 | 63 | ||
More Information
Set Membership: |
|

dcp2
Genomic Map