Assay Details
| Gene Symbol: mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362389.1 | NP_001349318.1 | 2 - 3 | 219 | 93 | |
| NM_001362390.1 | NP_001349319.1 | 2 - 3 | 179 | 93 | ||
| NM_200586.2 | NP_956880.1 | 2 - 3 | 175 | 93 | ||
| GenBank mRNA | AY337025.1 | - | 1 - 2 | 83 | 93 | |
| BC056727.1 | - | 2 - 3 | 141 | 93 | ||
| BC065862.1 | - | 2 - 3 | 133 | 93 | ||
| DR716512.1 | - | 2 - 3 | 167 | 93 | ||
| EH493828.1 | - | 2 - 3 | 191 | 93 | ||
More Information
Set Membership: |
|

mb
Genomic Map