Assay Details
| Gene Symbol: mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362389.1 | NP_001349318.1 | 3 - 4 | 437 | 69 | |
| NM_001362390.1 | NP_001349319.1 | 3 - 4 | 397 | 69 | ||
| NM_200586.2 | NP_956880.1 | 3 - 4 | 393 | 69 | ||
| GenBank mRNA | AY337025.1 | - | 2 - 3 | 301 | 69 | |
| BC056727.1 | - | 3 - 4 | 359 | 69 | ||
| BC065862.1 | - | 3 - 4 | 351 | 69 | ||
| DR716512.1 | - | 3 - 4 | 385 | 69 | ||
| EH493828.1 | - | 3 - 4 | 409 | 69 | ||
More Information
Set Membership: |
|

mb
Genomic Map