Assay Details
| Gene Symbol: gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213110.1 | NP_998275.1 | 6 - 7 | 464 | 102 | |
| GenBank mRNA | BC049318.1 | - | 6 - 7 | 464 | 102 | |
| BC075776.1 | - | 6 - 7 | 430 | 102 | ||
More Information
Set Membership: |
|

gmfb
Genomic Map