Assay Details
| Gene Symbol: morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017666.1 | NP_001017666.1 | 6 | 949 | 129 | |
| GenBank mRNA | BC093170.1 | - | 6 | 949 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

morn3| Gene Symbol: morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017666.1 | NP_001017666.1 | 6 | 949 | 129 | |
| GenBank mRNA | BC093170.1 | - | 6 | 949 | 129 | |
Set Membership: |
|