Assay Details
| Gene Symbol: adrb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089471.2 | NP_001082940.2 | 2 | 3632 | 108 | |
| GenBank mRNA | BC139539.1 | - | 3 | 3553 | 108 | |
More Information
Set Membership: |
|

adrb2b
Genomic Map