Assay Details
Gene Symbol: icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017618.1 | NP_001017618.1 | 5 | 1452 | 143 | |
GenBank mRNA | BC092830.1 | - | 5 | 1452 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
