Assay Details
| Gene Symbol: lurap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105708.1 | NP_001099178.1 | 3 | 1351 | 139 | |
| GenBank mRNA | BC151995.1 | - | 3 | 1351 | 139 | |
More Information
Set Membership: |
|

lurap1
Genomic Map