Assay Details
| Gene Symbol: cebpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131884.2 | NP_571959.2 | 1 | 1348 | 154 | |
| GenBank mRNA | AF306854.1 | - | 1 | 1327 | 154 | |
| BC049401.1 | - | 1 | 1348 | 154 | ||
| BC062523.1 | - | 1 | 1169 | 154 | ||
More Information
Set Membership: |
|

cebpb
Genomic Map