Assay Details
| Gene Symbol: znfl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194390.2 | NP_919371.2 | 2 | 184 | 67 | |
| GenBank mRNA | AB097828.1 | - | 2 | 154 | 67 | |
| BC092919.1 | - | 2 | 171 | 67 | ||
| BC164077.1 | - | 2 | 188 | 67 | ||
| EH504227.1 | - | 2 | 206 | 67 | ||
More Information
Set Membership: |
|

znfl2a
Genomic Map