Assay Details
| Gene Symbol: dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003442.1 | NP_001003442.1 | 6 - 7 | 536 | 75 | |
| GenBank mRNA | BC076064.1 | - | 6 - 7 | 536 | 75 | |
More Information
Set Membership: |
|

dnal1
Genomic Map