Assay Details
| Gene Symbol: cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007447.1 | NP_001007448.1 | 5 - 6 | 1919 | 69 | |
| GenBank mRNA | BC085392.1 | - | 5 - 6 | 1919 | 69 | |
| BC164562.1 | - | 4 - 5 | 1714 | 69 | ||
More Information
Set Membership: |
|

cpt2
Genomic Map