Assay Details
| Gene Symbol: fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013466.1 | NP_001013484.1 | 4 - 5 | 507 | 97 | |
| GenBank mRNA | BC091532.1 | - | 4 - 5 | 507 | 97 | |
More Information
Set Membership: |
|

fam221a
Genomic Map