Assay Details
| Gene Symbol: lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025472.1 | NP_001020643.1 | 9 - 10 | 1888 | 75 | |
| XM_005157631.4 | XP_005157688.1 | 10 - 11 | 1981 | 75 | ||
| XM_005157632.4 | XP_005157689.1 | 10 - 11 | 1966 | 75 | ||
| XM_005157633.4 | XP_005157690.1 | 9 - 10 | 1930 | 75 | ||
| GenBank mRNA | BC097149.1 | - | 9 - 10 | 1888 | 75 | |
More Information
Set Membership: |
|

lsr
Genomic Map