Assay Details
| Gene Symbol: hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100102.2 | NP_001093572.2 | 2 - 3 | 138 | 83 | |
| GenBank mRNA | EF158445.1 | - | 2 - 3 | 125 | 83 | |
| EH454362.1 | - | 2 - 3 | 124 | 83 | ||
More Information
Set Membership: |
|

hcst
Genomic Map