Assay Details
| Gene Symbol: npy1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102391.1 | NP_001095861.1 | 1 - 2 | 729 | 84 | |
| XM_005170925.3 | XP_005170982.1 | 2 - 3 | 1029 | 84 | ||
More Information
Set Membership: |
|

npy1r
Genomic Map