Assay Details
| Gene Symbol: yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002096.1 | NP_001002096.1 | 5 - 6 | 621 | 70 | |
| XM_005171830.1 | XP_005171887.1 | 5 - 6 | 445 | 70 | ||
| GenBank mRNA | BC071525.1 | - | 5 - 6 | 621 | 70 | |
More Information
Set Membership: |
|

yipf6
Genomic Map