Assay Details
| Gene Symbol: dhrs11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002143.2 | NP_001002143.2 | 5 - 6 | 701 | 65 | |
| GenBank mRNA | BC071398.1 | - | 5 - 6 | 694 | 65 | |
More Information
Set Membership: |
|

dhrs11b
Genomic Map