Assay Details
Gene Symbol: dhrs11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002143.2 | NP_001002143.2 | 6 - 7 | 769 | 105 | |
GenBank mRNA | BC071398.1 | - | 6 - 7 | 762 | 105 | |
EH518875.1 | - | 1 - 2 | 677 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
