Assay Details
Gene Symbol: shox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC107618.1 | - | 5 | 793 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
