Assay Details
Gene Symbol: st6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045370.1 | NP_001038835.1 | 3 - 4 | 237 | 75 | |
GenBank mRNA | AJ868430.1 | - | 1 - 2 | 152 | 75 | |
BC122199.1 | - | 3 - 4 | 237 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
