Assay Details
| Gene Symbol: nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312912.1 | NP_001299841.1 | 6 - 7 | 683 | 77 | |
| XM_017359027.1 | XP_017214516.1 | 5 - 6 | 472 | 77 | ||
| GenBank mRNA | BC122134.1 | - | 4 - 5 | 333 | 77 | |
| BC164740.1 | - | 3 - 4 | 278 | 77 | ||
More Information
Set Membership: |
|

nxt2
Genomic Map