Assay Details
| Gene Symbol: znf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077536.1 | NP_001071004.1 | 20 - 21 | 3269 | 71 | |
| GenBank mRNA | BC125817.1 | - | 20 - 21 | 3269 | 71 | |
More Information
Set Membership: |
|

znf185
Genomic Map