Assay Details
| Gene Symbol: opa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080166.1 | NP_001073635.1 | 1 - 2 | 165 | 104 | |
| GenBank mRNA | BC049503.1 | - | 1 - 2 | 165 | 104 | |
More Information
Set Membership: |
|

opa3
Genomic Map