Assay Details
| Gene Symbol: bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089463.1 | NP_001082932.1 | 3 - 4 | 339 | 82 | |
| XM_021467562.1 | XP_021323237.1 | 2 - 3 | 311 | 82 | ||
| GenBank mRNA | BC139669.1 | - | 3 - 4 | 339 | 82 | |
More Information
Set Membership: |
|

bbs10
Genomic Map