Assay Details
| Gene Symbol: p2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089527.1 | NP_001082996.1 | 2 - 3 | 291 | 72 | |
| XM_021480954.1 | XP_021336629.1 | 1 - 2 | 635 | 72 | ||
| GenBank mRNA | BC134131.1 | - | 2 - 3 | 291 | 72 | |
More Information
Set Membership: |
|

p2ry10
Genomic Map