Assay Details
| Gene Symbol: march8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161435.2 | NP_001154907.1 | 5 - 6 | 581 | 68 | |
| NM_001327919.1 | NP_001314848.1 | 6 - 7 | 1412 | 68 | ||
| XM_009307233.3 | XP_009305508.1 | 6 - 7 | 1406 | 68 | ||
| XM_009307235.3 | XP_009305510.1 | 6 - 7 | 1616 | 68 | ||
| XM_009307236.2 | XP_009305511.1 | 6 - 7 | 1613 | 68 | ||
| XM_009307237.3 | XP_009305512.1 | 5 - 6 | 1578 | 68 | ||
| XM_009307238.3 | XP_009305513.1 | 5 - 6 | 677 | 68 | ||
| XM_009307239.3 | XP_009305514.1 | 4 - 5 | 724 | 68 | ||
| GenBank mRNA | BC153940.1 | - | 4 - 5 | 536 | 68 | |
More Information
Set Membership: |
|

march8
Genomic Map