Assay Details
| Gene Symbol: dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113588.1 | NP_001107060.1 | 1 - 2 | 398 | 79 | |
| GenBank mRNA | BC154601.1 | - | 1 - 2 | 398 | 79 | |
| BC171432.1 | - | 1 - 2 | 353 | 79 | ||
| BC171434.1 | - | 1 - 2 | 353 | 79 | ||
More Information
Set Membership: |
|

dnajc18
Genomic Map