Assay Details
| Gene Symbol: tnmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114413.1 | NP_001107885.1 | 3 - 4 | 409 | 64 | |
| GenBank mRNA | BC155615.1 | - | 3 - 4 | 409 | 64 | |
More Information
Set Membership: |
|

tnmd
Genomic Map