Assay Details
Gene Symbol: mfsd4ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114416.1 | NP_001107888.1 | 1 - 2 | 284 | 98 | |
GenBank mRNA | BC155330.1 | - | 1 - 2 | 284 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
