Assay Details
Gene Symbol: mfsd4ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114416.1 | NP_001107888.1 | 9 - 10 | 1507 | 72 | |
GenBank mRNA | BC090783.1 | - | 2 - 3 | 166 | 72 | |
BC155330.1 | - | 9 - 10 | 1507 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
