Assay Details
| Gene Symbol: wnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201398.1 | NP_001188327.1 | 3 - 4 | 671 | 63 | |
| XM_005162223.4 | XP_005162280.1 | 4 - 5 | 1377 | 63 | ||
| GenBank mRNA | BC163003.1 | - | 3 - 4 | 671 | 63 | |
| BC163014.1 | - | 3 - 4 | 671 | 63 | ||
| KM655701.1 | - | 3 - 4 | 620 | 63 | ||
More Information
Set Membership: |
|

wnt1
Genomic Map