Assay Details
| Gene Symbol: npy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131074.2 | NP_571149.1 | 2 - 3 | 240 | 77 | |
| GenBank mRNA | BC162068.1 | - | 2 - 3 | 240 | 77 | |
| BC162071.1 | - | 2 - 3 | 240 | 77 | ||
More Information
Set Membership: |
|

npy
Genomic Map