Assay Details
| Gene Symbol: mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199871.1 | NP_956165.1 | 1 - 2 | 267 | 75 | |
| GenBank mRNA | BC045343.1 | - | 1 - 2 | 267 | 75 | |
| BC164484.1 | - | 1 - 2 | 109 | 75 | ||
More Information
Set Membership: |
|

mat1a
Genomic Map