Assay Details
| Gene Symbol: ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200107.2 | NP_956401.1 | 2 - 3 | 342 | 150 | |
| GenBank mRNA | BC053258.1 | - | 2 - 3 | 321 | 150 | |
| EH581922.1 | - | 2 - 3 | 359 | 150 | ||
| HM114348.1 | - | 2 - 3 | 262 | 150 | ||
More Information
Set Membership: |
|

ddit4
Genomic Map